ADVH14DBC120 AChemBlock

1-Boc-3-[(methylsulfonyl)oxy]piperidine

  • MDL number MFCD07787193
  • Cas Number : 129888-60-4
  • Molecular Weight : 279.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/1-boc-3-methylsulfonyl-oxy-piperidine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P41774
Availability details for
Quantity
  • Availability
  • Add to Cart