ADVH14DBC160 AChemBlock

tert-Butyl (3-hydrazino-3-oxopropyl)carbamate

  • MDL number MFCD08689453
  • Cas Number : 42116-56-3
  • Molecular Weight : 203.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-3-hydrazino-3-oxopropyl-carbamate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P38149
Availability details for
Quantity
  • Availability
  • Add to Cart