ADVH14DBC170 AChemBlock

2-Chloro-1-(4-methyl-1-piperazinyl)ethanone hydrochloride

  • MDL number MFCD04113883
  • Cas Number : 40340-73-6
  • Molecular Weight : 213.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/2-chloro-1-4-methyl-1-piperazinyl-ethanone-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P38238
Availability details for
Quantity
  • Availability
  • Add to Cart