ADVH14DBC173 AChemBlock

2-Methoxy-1-propanamine hydrochloride

  • MDL number MFCD08146657
  • Cas Number : 70807-90-8
  • Molecular Weight : 125.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/2-methoxy-1-propanamine-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P36510
Availability details for
Quantity
  • Availability
  • Add to Cart