ADVH14DBC199 AChemBlock

2-Methyl-benzenepropanamine

  • MDL number MFCD07374073
  • Cas Number : 76293-74-8
  • Molecular Weight : 149.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/2-methyl-benzenepropanamine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P36201
Availability details for
Quantity
  • Availability
  • Add to Cart