ADVH14DBC1B3 AChemBlock

3-Fluoro-benzenebutanamine

  • MDL number MFCD19677879
  • Cas Number : 1339855-93-4
  • Molecular Weight : 167.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/3-fluoro-benzenebutanamine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P41608
Availability details for
Quantity
  • Availability
  • Add to Cart