ADVH14DBC1C2 AChemBlock

4-(1-Pyrrolidinyl)-cyclohexanamine dihydrochloride

  • MDL number MFCD30011360
  • Cas Number : 1001347-35-8
  • Molecular Weight : 241.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/4-1-pyrrolidinyl-cyclohexanamine-dihydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P43133
Availability details for
Quantity
  • Availability
  • Add to Cart