ADVH14DBC1C9 AChemBlock

1,2,3,4-Tetrahydro-9-aminoacridine

  • MDL number MFCD00046923
  • Cas Number : 321-64-2
  • Molecular Weight : 198.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/1-2-3-4-tetrahydro-9-aminoacridine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P38835
Availability details for
Quantity
  • Availability
  • Add to Cart