ADVH14DBC1D9 AChemBlock

Methyl 2,3-dioxo-1H-indoline-5-carboxylate

  • MDL number MFCD11045030
  • Cas Number : 101460-85-9
  • Molecular Weight : 205.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/methyl-2-3-dioxo-1h-indoline-5-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P43068
Availability details for
Quantity
  • Availability
  • Add to Cart