ADVH14DBC1EB AChemBlock

4-Chloro-7-nitroquinazoline

  • MDL number MFCD00033667
  • Cas Number : 19815-17-9
  • Molecular Weight : 209.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/4-chloro-7-nitroquinazoline.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P40069
Availability details for
Quantity
  • Availability
  • Add to Cart