ADVH14DBC1EE AChemBlock

1H-Pyrazole-3-carboxylic acid methyl ester

  • MDL number MFCD00649381
  • Cas Number : 15366-34-4
  • Molecular Weight : 126.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/1h-pyrazole-3-carboxylic-acid-methyl-ester.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P40994
Availability details for
Quantity
  • Availability
  • Add to Cart