ADVH14DBC201 AChemBlock

1-Trityl-imidazole-4-carboxaldehyde

  • MDL number MFCD02179554
  • Cas Number : 33016-47-6
  • Molecular Weight : 338.41

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/1-trityl-imidazole-4-carboxaldehyde.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P38789
Availability details for
Quantity
  • Availability
  • Add to Cart