ADVH14DBC20F AChemBlock

6-Bromochroman

  • MDL number MFCD10698725
  • Cas Number : 3875-78-3
  • Molecular Weight : 213.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/6-bromochroman.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P38383
Availability details for
Quantity
  • Availability
  • Add to Cart