ADVH14DBC21A AChemBlock

1-Boc-3-iodoindazole

  • MDL number MFCD09835520
  • Cas Number : 290368-00-2
  • Molecular Weight : 344.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/1-boc-3-iodoindazole.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P39029
Availability details for
Quantity
  • Availability
  • Add to Cart