ADVH14DBC238 AChemBlock

4-Iodo-1-tritylimidazole

  • MDL number MFCD02179542
  • Cas Number : 96797-15-8
  • Molecular Weight : 436.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/4-iodo-1-tritylimidazole.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P34785
Availability details for
Quantity
  • Availability
  • Add to Cart