ADVH14DBC23E AChemBlock

5-Methoxy-2,3-dihydro-1H-indole

  • MDL number MFCD07368037
  • Cas Number : 21857-45-4
  • Molecular Weight : 149.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/5-methoxy-2-3-dihydro-1h-indole.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P39671
Availability details for
Quantity
  • Availability
  • Add to Cart