ADVH14DBC265 AChemBlock

5-Methoxy-2-benzofurancarboxylic acid

  • MDL number MFCD00079771
  • Cas Number : 10242-08-7
  • Molecular Weight : 192.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/5-methoxy-2-benzofurancarboxylic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P42968
Availability details for
Quantity
  • Availability
  • Add to Cart