ADVH14DBC298 AChemBlock

b-Amino-2-methoxybenzeneethanol

  • MDL number MFCD08752814
  • Cas Number : 108343-90-4
  • Molecular Weight : 167.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/b-amino-2-methoxybenzeneethanol.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P42721
Availability details for
Quantity
  • Availability
  • Add to Cart