ADVH14DBC2AF AChemBlock

N-Boc-(R)-2-hydroxypropanamine

  • MDL number MFCD04974338
  • Cas Number : 119768-44-4
  • Molecular Weight : 175.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/n-boc-r-2-hydroxypropanamine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P42353
Availability details for
Quantity
  • Availability
  • Add to Cart