ADVH14DBC2B7 AChemBlock

(S)-tert-Butyl 3-(Fmoc-amino)-4-hydroxybutanoate

  • MDL number MFCD01861297
  • Cas Number : 133565-45-4
  • Molecular Weight : 397.48

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/s-tert-butyl-3-fmoc-amino-4-hydroxybutanoate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P41629
Availability details for
Quantity
  • Availability
  • Add to Cart