ADVH14DBC2DB AChemBlock

1-(2-Hydroxy-2-phenylethyl)pyrrolidine

  • MDL number MFCD02129879
  • Cas Number : 5407-61-4
  • Molecular Weight : 191.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/1-2-hydroxy-2-phenylethyl-pyrrolidine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P37545
Availability details for
Quantity
  • Availability
  • Add to Cart