ADVH14DBC2F9 AChemBlock

1-Piperidinebutanamine

  • MDL number MFCD03727285
  • Cas Number : 74247-30-6
  • Molecular Weight : 156.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/1-piperidinebutanamine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P36333
Availability details for
Quantity
  • Availability
  • Add to Cart