ADVH14DBC302 AChemBlock

N2,N2,2-trimethylpropane-1,2-diamine dihydrochloride

  • MDL number MFCD09836383
  • Cas Number : 76936-44-2
  • Molecular Weight : 189.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/n2-n2-2-trimethylpropane-1-2-diamine-dihydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P36156
Availability details for
Quantity
  • Availability
  • Add to Cart