ADVH14DBC325 AChemBlock

N-[4-(Aminomethyl)phenyl]acetamide hydrochloride

  • MDL number MFCD06656043
  • Cas Number : 25027-73-0
  • Molecular Weight : 200.67

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/n-4-aminomethyl-phenyl-acetamide-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P39368
Availability details for
Quantity
  • Availability
  • Add to Cart