ADVH14DBC32C AChemBlock

N-Methyl-a-phenyl-1-pyrrolidineethanamine

  • MDL number MFCD01862531
  • Cas Number : 130110-09-7
  • Molecular Weight : 204.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/n-methyl-a-phenyl-1-pyrrolidineethanamine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P41767
Availability details for
Quantity
  • Availability
  • Add to Cart