ADVH14DBC373 AChemBlock

Fmoc-4-[2-(Boc-amino)ethoxy]-L-phenylalanine

  • MDL number MFCD07781260
  • Cas Number : 1013883-02-7
  • Molecular Weight : 546.63

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/fmoc-4-2-boc-amino-ethoxy-l-phenylalanine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P43075
Availability details for
Quantity
  • Availability
  • Add to Cart