ADVH14DBC42D AChemBlock

Methyl 1-Aminocyclohexanecarboxylate hydrochloride

  • MDL number MFCD09264353
  • Cas Number : 37993-32-1
  • Molecular Weight : 193.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/methyl-1-aminocyclohexanecarboxylate-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P38427
Availability details for
Quantity
  • Availability
  • Add to Cart