ADVH14DBC461 AChemBlock

2-[(Fmoc-amino)oxy]acetic acid

  • MDL number MFCD01076133
  • Cas Number : 123106-21-8
  • Molecular Weight : 313.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/2-fmoc-amino-oxy-acetic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P42137
Availability details for
Quantity
  • Availability
  • Add to Cart