ADVH14DBC477 AChemBlock

(S)-2-(Boc-amino)-4-iodobutyric acid methyl ester

  • MDL number MFCD16294352
  • Cas Number : 101650-14-0
  • Molecular Weight : 343.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/s-2-boc-amino-4-iodobutyric-acid-methyl-ester.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P43056
Availability details for
Quantity
  • Availability
  • Add to Cart