ADVH14DBC487 AChemBlock

N-Fmoc-6-fluoro-L-tryptophan

  • MDL number MFCD29917530
  • Cas Number : 908847-01-8
  • Molecular Weight : 444.47

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/n-fmoc-6-fluoro-l-tryptophan.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P35213
Availability details for
Quantity
  • Availability
  • Add to Cart