ADVH14DBC499 AChemBlock

a-(Dibutylamino)-4-methoxy-benzeneacetamide

  • MDL number MFCD00865229
  • Cas Number : 519-88-0
  • Molecular Weight : 292.42

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/a-dibutylamino-4-methoxy-benzeneacetamide.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P37669
Availability details for
Quantity
  • Availability
  • Add to Cart