ADVH14DBC51A AChemBlock

Boc-(R)-2-amino-2-(4-hydroxyphenyl)acetic acid

  • MDL number MFCD01861312
  • Cas Number : 27460-85-1
  • Molecular Weight : 267.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/boc-r-2-amino-2-4-hydroxyphenyl-acetic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P39129
Availability details for
Quantity
  • Availability
  • Add to Cart