ADVH14DBC525 AChemBlock

Boc-(R)-2-amino-2-(3-fluorophenyl)acetic acid

  • MDL number MFCD07371709
  • Cas Number : 209680-91-1
  • Molecular Weight : 269.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/boc-r-2-amino-2-3-fluorophenyl-acetic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P39836
Availability details for
Quantity
  • Availability
  • Add to Cart