ADVH14DBC526 AChemBlock

3-Amino-benzeneacetic acid ethyl ester hydrochloride

  • MDL number MFCD27500662
  • Cas Number : 52273-79-7
  • Molecular Weight : 215.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/3-amino-benzeneacetic-acid-ethyl-ester-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P37644
Availability details for
Quantity
  • Availability
  • Add to Cart