ADVH14DBC54F AChemBlock

(S)-3-Amino-4-(4-chlorophenyl)butyric acid

  • MDL number MFCD04113703
  • Cas Number : 270596-41-3
  • Molecular Weight : 213.67

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/s-3-amino-4-4-chlorophenyl-butyric-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P39154
Availability details for
Quantity
  • Availability
  • Add to Cart