ADVH14DBC56F AChemBlock

(R)-a-(Fmoc-amino)cyclobutaneacetic acid

  • MDL number MFCD27951989
  • Cas Number : 923012-41-3
  • Molecular Weight : 351.41

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/r-a-fmoc-amino-cyclobutaneacetic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P35072
Availability details for
Quantity
  • Availability
  • Add to Cart