ADVH14DBC572 AChemBlock

4'-Amino-biphenyl-4-acetic acid

  • MDL number MFCD05980686
  • Cas Number : 1123169-05-0
  • Molecular Weight : 227.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/4-amino-biphenyl-4-acetic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P42607
Availability details for
Quantity
  • Availability
  • Add to Cart