ADVH14DBC575 AChemBlock

(S)-b-Aminobenzenebutanoic acid

  • MDL number MFCD03428441
  • Cas Number : 26250-87-3
  • Molecular Weight : 179.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/s-b-aminobenzenebutanoic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P39247
Availability details for
Quantity
  • Availability
  • Add to Cart