ADVH14DBC576 AChemBlock

(R)-b-Aminobenzenebutanoic acid

  • MDL number MFCD11615764
  • Cas Number : 131270-08-1
  • Molecular Weight : 179.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/r-b-aminobenzenebutanoic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P41730
Availability details for
Quantity
  • Availability
  • Add to Cart