ADVH14DBC5AE AChemBlock

D-Tic-OEt.hydrochloride

  • MDL number MFCD10565741
  • Cas Number : 41220-49-9
  • Molecular Weight : 241.72

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/d-tic-oet-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P38179
Availability details for
Quantity
  • Availability
  • Add to Cart