ADVH14DBC607 AChemBlock

N-Boc-3-(4-methylbenzyl)piperidine-3-carboxylic acid

  • MDL number MFCD03840050
  • Cas Number : 887344-23-2
  • Molecular Weight : 333.43

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/n-boc-3-4-methylbenzyl-piperidine-3-carboxylic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P35402
Availability details for
Quantity
  • Availability
  • Add to Cart