ADVH14DBC60C AChemBlock

4-(Aminomethyl)benzoic acid tert-butyl ester

  • MDL number MFCD04973451
  • Cas Number : 107045-28-3
  • Molecular Weight : 207.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/4-aminomethyl-benzoic-acid-tert-butyl-ester.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P42759
Availability details for
Quantity
  • Availability
  • Add to Cart