ADVH14DBC640 AChemBlock

1-Amino-3-benzyloxycyclobutane-1-carboxylic acid

  • MDL number MFCD07644566
  • Cas Number : 191110-50-6
  • Molecular Weight : 221.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/1-amino-3-benzyloxycyclobutane-1-carboxylic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P34710
Availability details for
Quantity
  • Availability
  • Add to Cart