ADVH14DBC6C9 AChemBlock

(S)-2-Methylsuccinic acid

  • MDL number MFCD00192321
  • Cas Number : 2174-58-5
  • Molecular Weight : 132.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/s-2-methylsuccinic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P39685
Availability details for
Quantity
  • Availability
  • Add to Cart