ADVH14DBC6DA AChemBlock

Ethyl 2,3-dihydroxybenzeneacetate

  • MDL number MFCD29917870
  • Cas Number : 342623-35-2
  • Molecular Weight : 196.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/ethyl-2-3-dihydroxybenzeneacetate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P38689
Availability details for
Quantity
  • Availability
  • Add to Cart