ADVH14DBC735 AChemBlock

2'-Methylbiphenyl-3-carboxylic acid

  • MDL number MFCD03424589
  • Cas Number : 168618-44-8
  • Molecular Weight : 212.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/2-methylbiphenyl-3-carboxylic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P40659
Availability details for
Quantity
  • Availability
  • Add to Cart