ADVH14DBC73D AChemBlock

2'-Fluoro-4-biphenylcarboxylic acid

  • MDL number MFCD00278732
  • Cas Number : 365-12-8
  • Molecular Weight : 216.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/2-fluoro-4-biphenylcarboxylic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P38516
Availability details for
Quantity
  • Availability
  • Add to Cart