ADVH14DBC748 AChemBlock

4'-Acetyl-4-biphenylcarboxylic acid

  • MDL number MFCD00230828
  • Cas Number : 114691-92-8
  • Molecular Weight : 240.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/4-acetyl-4-biphenylcarboxylic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P42546
Availability details for
Quantity
  • Availability
  • Add to Cart