ADVH14DBC788 AChemBlock

2-Methyl-2-phenyl-1,3-propanediol

  • MDL number MFCD00130135
  • Cas Number : 24765-53-5
  • Molecular Weight : 166.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/2-methyl-2-phenyl-1-3-propanediol.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P39384
Availability details for
Quantity
  • Availability
  • Add to Cart