ADVH14DBC7C2 AChemBlock

1-Ethynyl-2,4-dimethyl-benzene

  • MDL number MFCD08703476
  • Cas Number : 16017-30-4
  • Molecular Weight : 130.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/1-ethynyl-2-4-dimethyl-benzene.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P40859
Availability details for
Quantity
  • Availability
  • Add to Cart